Learn More
ACCU-SCOPE™ Excelis™ HD Lite Camera
SDP
Catalog No. 01189221 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
12 V
Catalog No. Voltage
01-189-221 12 V
1 options
Catalog No. 01-189-221 Supplier ACCU-SCOPE™ Supplier No. AU300HD
Add to Cart
Add to Cart
The Excelis™ 1080P HD is a high-fidelity color microscopy camera delivers true, real-time 1080P HD images for a wide variety of scientific, education and industrial applications. On-board software allows operation without a PC or use with CaptaVision+ v3.0 advanced imaging software.
  • HD live imaging for scientific and industrial applications. Capture 5 MP still images.
  • View and capture images without a PC (HDMI monitor required).
  • Built-in mouse control software for HDMI viewing and recording without a computer.
  • CaptaVision+ v3.0 Imaging Software (included) extends the capabilities of the Excelis™ HD with advanced image processing and measurement features (requires PC, not included)
  • Standard c-mount easily adapts to most microscopes, 0.35x camera adapter recommended.
  • 1-year warranty
  • In the box: Excelis™ HD Camera, HDMI Cable, USB 2.0 Cable, 12 V Power Adapter, 8 GB SD Card, USB Mouse, CaptaVision+ v3.0 PC Imaging Software
Specifications
Type USB Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 12 V