Learn More
Supply Solutions EX-CELLAntifoam, gamma irradiated, 1 Liter
Catalog No. NC1927382
Encompass_Preferred
EX-CELLANTIFOAM is a USP Grade non-ionic emulsion (simethicone) designed for use in the pharmaceutical and veterinary biological industries to aid in the control of foaming typically associated with the use of culture medium in bioreactors.
Catalog No. NC1927382 Supplier Supply Solutions Supplier No. SIGMA 59920C1B
EX-CELLANTIFOAM is a USP Grade non-ionic emulsion (simethicone) designed for use in the pharmaceutical and veterinary biological industries to aid in the control of foaming typically associated with the use of culture medium in bioreactors.