Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Excell Plus Fixative
SDP

Catalog No. 21033370
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-033-370 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-370 Supplier StatLab Supplier No. FXEXPGAL
Add to Cart
Add to Cart

A low-hazard tissue fixative alternative to 10% formalin

  • ExCell Plus has a pleasant aroma and contains No Heavy Metals
  • This fixative firms tissue and penetrates most tissue specimens as fast as 10% Formalin
  • Enhances microtome sectioning for H-E stains, Special Stains, Immuno-Stains, Flowcytometry, DNA Analysis, and Fluorescence In Situ Hybridization (FISH)
  • ExCell Plus is virucidal biocidal and fungicidal

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.