Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab ExCell Plus Tissue Fixative
SDP

Catalog No. 22021428
Change view
Click to view available options
Quantity:
20 mL
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-428 20 mL
22-021-414 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-428 Supplier StatLab Supplier No. FXEXP20CS
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab ExCell Plus Tissue Fixative - a low-hazard alternative to 10% formalin

  • The only tissue fixative that actually smells pleasant
  • ExCell Plus contains No Heavy Metals
  • This amazing tissue fixative firms tissue and penetrates most tissue specimens as fast as 10% Formalin
  • ExCell Plus enhances microtome sectioning, H and E stains, Special Stains, Immuno-Stains, Flowcytometry, DNA Analysis, and Fluorescence In Situ Hybridization (FISH)
  • ExCell Plus is virucidal, biocidal and fungicidal

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 20 mL
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.