Learn More
Epredia™ Excelsior™ AS Tissue Processor
Catalog No. A82300001ER Shop All Epredia Products
Change view
Click to view available options
Includes:
Line Cond Mcrtme, Blade Car ER
Line Conditioner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
A82300001ER Line Cond Mcrtme, Blade Car ER
A82300001A Line Conditioner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A82300001ER Supplier Epredia™ Supplier No. A82300001ER
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Reduce reagent costs while enhancing workflow and tissue quality with the Epredia™ Excelsior™ AS Tissue Processor.
Enhanced tissue quality
  • Cassette baskets are gently rotated inside the circular chamber, providing more effective agitation than an impeller
  • Reagents can be pre-heated to up to 35°C, minimizing tissue shock and facilitating reagent penetration
Significant reagent cost savings
  • Alcohol quality measurement enables you to extend your reagent life and provides significant cost savings
  • Automatic reagent rotation further extends reagent life using a proven system
Streamline workflow and increase productivity
  • Reduce maintenance time by replacing one bottle of each reagent, rather than all of them
  • Draw new reagents directly into the processor from your supplier's bottles—no pouring, no mess, and no fumes
  • Run rapid biopsies during the day and conventional programs overnight
  • Compatible with IPA xylene-free processes as well as conventional reagents
Safe for the tissue — safe for the operator
  • When the chamber is opened, a fan pulls fumes through a filter to protect users from formaldehyde exposure
  • Fumes from the cabinet are also extracted through filters to protect users from alcohol, xylene, and formaldehyde
  • Waste paraffin can be removed by simply discarding a plastic tray—no paraffin spillage and burns
  • Power outages are no longer a concern; Excelsior keeps your tissue safe with battery backup
So simple to learn and operate
  • The all-new touch-screen interface is intuitive and easy-to-learn
  • With intelligent software, it's easier to avoid mistakes that affect tissue quality
Engineered for reliability and peace of mind
  • The Excelsior AS Tissue Processor reflects engineering enhancements based on more than 10 years of service history
Infomatics to integrate the laboratory and simplify reporting
  • A full range of reports is available for routine logging, quality assessment, statutory reporting and trouble shooting
  • Reports are easily downloaded via a USB drive
  • Messages can be sent automatically to the laboratory information system
  • Remote alarms ensure that any problems are immediately addressed
  • Epredia™ NetMon™ II monitoring system allows remote diagnostics over a HIPPA compliant, secure internet connection for immediate identification and resolution
Specifications
Product Type Tissue Processor
Includes Line Cond Mcrtme, Blade Car ER
Electrical Requirements 1300 VA (max.), 300 VA (typical)
Frequency 50/60 Hz
Voltage 100/240 V