Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Excelsior™ AS Tissue Processor Accessories

Catalog No. B35210007 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Charcoal Filter
Organized Basket
Potassium Permanganate Filter
Storage Bottle
Waste Wax Drawer Kit
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
B35210007 Basket Carrying Container
A78410024 Air System Duct Adapter Kit
A78410026 Storage Bottle
A78410027 Waste Wax Drawer Kit
74-112-58 Charcoal Filter
B99-906-12CS Potassium Permanganate Filter
A78410025 Organized Basket
A82310038 Organized Basket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. B35210007 Supplier Epredia™ Supplier No. B35210007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.

Product Features:
  • Microwaveable design allowing pre-processing of tissue
  • Organized capacity of 300 cassettes in six basket-halves

Ordering Alerts:

Basket halves only match with each other but can be mixed with 222 cassette baskets or SecureSette Excelsior ES baskets

Includes:

Six 50-cassette basket halves and user guide and loading instructions

Compatible with:

Excelsior ES Tissue Processor

Specifications

Product Type Basket Carrying Container
For Use With (Equipment) Excelsior AS Tissue Processor
Material Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.