Learn More
Electron Microscopy Sciences DiATOME Histo Cryo Diamond Knife Exchange Dry 6.0 mm
SDP
Catalog No. 501929856
Encompass
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission. Exchange any diamond knife for a new DiATOME Histo Cryo Diamond Knife. It does not matter what size or boat style that is traded in.
Size: 6.0 mm
Knife angle: 45°
Catalog No. 50-192-9856 Supplier Electron Microscopy Sciences Supplier No. 60HDCE
These diamond knives are for use at cryo temperatures. The knives have a 45 degree angle and section thickness range 0.2 - 5 micron. The histo-cryo knives are delivered in boats for wet cryo sectioning and triangular holders for dry cryo sectioning, made of a special copper/nickel alloy, which guarantees the best heat/cold transmission. Exchange any diamond knife for a new DiATOME Histo Cryo Diamond Knife. It does not matter what size or boat style that is traded in.
Size: 6.0 mm
Knife angle: 45°