Learn More
Metrohm USA EXCHANGE UNIT 50ML
SDP
Supplier:  Metrohm USA 63026250
Encompass
Exchange unit with integrated data chip with 50 mL glass cylinder and light protection. PCTFE/PTFE flat stopcock, FEP tubing connection, antidiffusion tip and standard amber glass reagent bottle.
Catalog No. NC0246298