Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSI™ Exchangeable Unit for Volitale Liquids

Catalog No. 15177866 Shop All Yellow Springs Instrument (YSI) Products
Change view
Click to view available options
Volume (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15-177-866 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-177-866 Supplier YSI™ Supplier No. 285220360
Add to Cart
Add to Cart

Intelligent exchangeable heads for the piston burette TITRONIC™ 500 and the titrators TitroLine™ 7000, 7500, 7750 und 7800.

Intelligent exchangeable heads for the piston burette TITRONIC™ 500 and the titrators TitroLine™ 7000, 7500, 7750 und 7800.

  • Size options of 5, 10, 20 und 50 ml
  • Compact, space saving footprint
  • Easy to exchange
  • All relevant reagent and unit data are stored in the integrated RFID-chip including:
    - Burette size (ml)
    - Reagent/titrant name
    - Titrant concentration or titer value of solution
    - Date of manufacture or expiration date of the reagent.
    ISO 8655 Certified (“C” certificate included)

Scope of delivery:

5, 10 , 20 or 50 ml burette, with integrated chip for reagent data, with 1 L brown glass bottle for titrant, GL 45 and S 40-bottle adapter, tubes, drip tube and titration tip

TRUSTED_SUSTAINABILITY

Specifications

Type Intelligent Exchangeable Unit for TitroLine 7000+ and TITRONIC 500
Volume (Metric) 50 mL
Volume (Metric) Sample 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.