Learn More
Med Vet International Excision Rasp Excision Arthoplasty, J0209, 6.5in, Each
Catalog No. 5000585559
Encompass_Preferred
The fine head on the rasp allows easy removal of bone spurs or rough edges in a femoral head excision. The rasp cuts only on the backward stroke.
Catalog No. 50-005-85559 Supplier Med Vet International Supplier No. JOR209
The fine head on the rasp allows easy removal of bone spurs or rough edges in a femoral head excision. The rasp cuts only on the backward stroke.