Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NIGHTSEA™ Stereo Fluorescence Add-On Light and Filter Set, Cyan
SDP

Catalog No. 501902444
Encompass_Preferred

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing a kit that consists of a light head, barrier filter, and viewing shield. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. Sets are named and color coded for the color of the excitation light, not the color of the emitted fluorescence.
Color: Cyan
Excitation: 490-515nm
Emission filter: 550 nm longpass
Fluorophores: YFP, Venus, Lucifer Yellow

Catalog No. 50-190-2444 Supplier Electron Microscopy Sciences Supplier No. SFALFSCY
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing a kit that consists of a light head, barrier filter, and viewing shield. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. Sets are named and color coded for the color of the excitation light, not the color of the emitted fluorescence.
Color: Cyan
Excitation: 490-515nm
Emission filter: 550 nm longpass
Fluorophores: YFP, Venus, Lucifer Yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.