Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences NIGHTSEA™ Stereo Fluorescence Add-On Light and Filter Set, Green-Only
SDP

Catalog No. 501931674
Encompass_Preferred

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing a kit that consists of a light head, barrier filter, and viewing shield. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. Sets are named and color coded for the color of the excitation light, not the color of the emitted fluorescence.
Color: Green-Only
Excitation: 440-460nm
Emission filter: 500-560 nm bandpass
Fluorophores: GFP, eGFP, fluorescein

Catalog No. 50-193-1674 Supplier Electron Microscopy Sciences Supplier No. SFALFSRBGO
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Additional combinations excitation/ emission wavelength combination can be added NIGHTSEA™ Stereo Microscope Fluorescence Viewing system by purchasing a kit that consists of a light head, barrier filter, and viewing shield. These three elements can be removed and replaced in seconds, and color coding ensures that you are using the right combination. Sets are named and color coded for the color of the excitation light, not the color of the emitted fluorescence.
Color: Green-Only
Excitation: 440-460nm
Emission filter: 500-560 nm bandpass
Fluorophores: GFP, eGFP, fluorescein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.