Learn More
NEOGENESTAR LLC ExDeep 24 Well Plate for 16mL, 25 plates/case
Catalog No. NC1831973
Encompass_Preferred
NeoGeneStar ExDeep 24 Well Plate for 16mL. 25 plates per case. Made from polypropylene. Sterile, Individually wrapped, perfect for culturing suspension cells. Perfect solution for automation platform
Catalog No. NC1831973 Supplier NEOGENESTAR LLC Supplier No. NGS 24 WELL 16 ML
NeoGeneStar ExDeep 24 Well Plate for 16mL. 25 plates per case. Made from polypropylene. Sterile, Individually wrapped, perfect for culturing suspension cells. Perfect solution for automation platform