Learn More
Exel International 20 to 25cc Syringes
Description
Latex free;Sterile;Non-toxic;Pyrogen free;Positive plunger stop to prevent spillage when drawing;Smooth plunger motion;Polypropylene construction.
Specifications
Specifications
| Certifications/Compliance | Pyrogen Free |
| Material | Polypropylene |
| Needle Gauge | No Needle Included |
| Latex Free | Yes |
| Product Type | Syringe |
| Volume (Metric) | 20 mL |
| Connection Type | Leur Slip |
| Graduated | Yes |
| Sterility | Sterile |