Learn More
Exel International 30 to 35cc Syringes
Catalog No. 1484129
Change view
Click to view available options
Connection Type:
Catheter
Leur Slip
Luer Lock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
14-841-29 Catheter
14-841-28 Leur Slip
14-841-27 Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-841-29 Supplier Exel International Supplier No. 26292
Add to Cart
Add to Cart
This item is not returnable. View return policy
Latex free;Sterile;Non-toxic;Pyrogen free;Positive plunger stop to prevent spillage when drawing;Smooth plunger motion;Polypropylene construction.
Specifications
Certifications/Compliance Pyrogen Free
Material Polypropylene
Latex Free Yes
Product Type Syringe
Volume (Metric) 30 mL
Connection Type Catheter
Graduated Yes
Sterility Sterile