Learn More
Exel International 5 to 6cc Syringes
Description
- Latex free
- Sterile
- Non-toxic
- Pyrogen free
- Positive plunger stop to prevent spillage when drawing
- Smooth plunger motion
- Polypropylene construction
Specifications
Specifications
| Certifications/Compliance | Pyrogen Free |
| Material | Polypropylene |
| For Use With (Application) | For general purpose |
| Latex Free | Yes |
| Product Type | Syringe |
| Volume (Metric) | 5 to 6 mL |
| Connection Type | Luer Lock |
| Graduated | Yes |
| Sterility | Sterile |