Learn More
Exel International 5 to 6cc Syringes
Catalog No. 1484147
Change view
Click to view available options
Connection Type:
Leur Slip
Luer Lock
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
14-841-47 Leur Slip
14-841-46 Luer Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-841-47 Supplier Exel International Supplier No. 26231
Add to Cart
Add to Cart
This item is not returnable. View return policy
  • Latex free
  • Sterile
  • Non-toxic
  • Pyrogen free
  • Positive plunger stop to prevent spillage when drawing
  • Smooth plunger motion
  • Polypropylene construction
Specifications
Certifications/Compliance Pyrogen Free
Latex Free Yes
Product Type Syringe
Volume (Metric) 5 to 6 mL
Connection Type Leur Slip
Sterility Sterile