Learn More
Exel International Multi-Sample Blood Draw Needles
Description
Sharp and smooth edges make penetration painless for patients and provide easy connection to rubber stoppers
Specifications
Specifications
| Product Type | Multi-sample Blood Draw Needle |
| Sterility | Sterile |
| Color | Yellow |
| For Use With (Application) | Provides several samples to be taken with a single puncture, make penetration painless for patients and provides easy connection to rubber stoppers |
| Gauge | 20 G |
| Includes | Sharp and Smooth edge |
| Latex Free | Yes |
| Length (English) | 1.5 in. |
| Length (Metric) | 3.8 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.