Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exel International Safety AV Fistula

Catalog No. 1484048
Change view
Click to view available options
Needle Gauge:
15 G
16 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
14-840-48 16 G
14-840-47 15 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-840-48 Supplier Exel International Supplier No. 27786

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Material Siliconized
Needle Gauge 16 G
For Use With (Application) Reduces possibility of accidental needle sticks, allow maximum blood flow
Includes Backeye, 12 in. tubing, clamp and green fixed wing, ultra-thin wall needle, luer cap
Latex Free Yes
Length (English) 1 in.
Product Type Secure Touch™ Av Fistula
Quantity 50 Box
Needle Point Style Ultra-sharp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.