Learn More
Exel International Tuberculin Syringes
Catalog No. 1484053
Change view
Click to view available options
Needle Gauge:
25 G
26 G
27 G
No Needle Included
Connection Type:
Fixed
Luer Tip
Removable
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Connection Type
14-840-53 No Needle Included Luer Tip
14-840-50 27 G Removable
14-840-51 26 G Removable
14-841-34 25 G Fixed
14-840-52 25 G Removable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-840-53 Supplier Exel International Supplier No. 26048
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
  • Latex free
  • Sterile
  • Non-toxic
  • Non pyrogenic
  • Reduced dead space for reduced medication waste
  • Smooth plunger action for greater filling accuracy
Specifications
Certifications/Compliance Pyrogen Free
Material Polypropylene
Needle Gauge No Needle Included
For Use With (Application) Positive plunger stop to prevent spillage when drawing, smooth plunger action for greater filling accuracy, non-toxic, pyrogen free
Latex Free Yes
Product Type Tuberculin Syringe
Volume (Metric) 1 mL
Connection Type Luer Tip
Sterility Sterile