Learn More
Medchemexpress LLC HY-13632 10mg Medchemexpress, Exemestane CAS:107868-30-4 Purity:>98%
SDP
Catalog No. 502081436
Encompass_Preferred
Medchemexpress, HY-13632 10mg Exemestane CAS:107868-30-4 Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1436 Supplier Medchemexpress LLC Supplier No. HY1363210MG
Medchemexpress, HY-13632 10mg Exemestane CAS:107868-30-4 Exemestane (FCE 24304) is a selective, irreversible and orally active steroidal aromatase inhibitor with IC50s of 30 nM and 40 nM for human placental and rat ovarian aromatase, respectively. Exemestane can be used for hormone-dependent breast cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.