Learn More
Selleck Chemical LLC Exemestane 50mg 107868-30-4 FCE24304, PNU155971
SDP
Supplier:  Selleck Chemical LLC S119650MG
Encompass_Preferred
Exemestane (FCE24304, PNU155971) is an aromatase inhibitor, inhibits human placental and rat ovarian aromatase with IC50 of 30 nM and 40 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-762-1