Learn More
Labelmaster Inc Exempt Specimen Label, Human 1000/roll
SDP
Catalog No. NC9509656
Encompass_Preferred
Labelmaster's Exempt Human Specimen Labels can be used for shipping human medical specimens. These paper labels comply with the requirements in International Air Transport, ICAO and DOT for marking the package orientation. The bright red label surface has outstanding chemical and abrasion resistance, and the backing offers strong adhesion to paint, steel, fiber and plastic surfaces. These labels keep you in regulatory compliance, measure 2 1/4 x 1 3/8 in. and are sold in rolls of 1000.
Catalog No. NC9509656 Supplier Labelmaster Inc Supplier No. L358
Labelmaster's Exempt Human Specimen Labels can be used for shipping human medical specimens. These paper labels comply with the requirements in International Air Transport, ICAO and DOT for marking the package orientation. The bright red label surface has outstanding chemical and abrasion resistance, and the backing offers strong adhesion to paint, steel, fiber and plastic surfaces. These labels keep you in regulatory compliance, measure 2 1/4 x 1 3/8 in. and are sold in rolls of 1000.