Learn More
CPC Scientific H-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific EXED002A
Encompass
Exendin (9-39) is a selective and potent against glucagon-like peptide 1 (GLP-1) and inhibits (Kd = 1.7 nm) cAMP production caused by Gastric Inhibitory Peptide (GIP). Exendin (9-39) is involved in appetite modulation by reducing insulin secretion and helps prevent sharp falls in glucose levels. This peptide has been studied for its? effects on the permeability of basal microvascular.SEQUENCE: H-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: DLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C149H234N40O47SMOLECULAR WEIGHT: 3369.79STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [133514-43-9]SYNONYMS: Exendin-(9-39)RESEARCH AREA: DiabetesREFERENCES:Greig, NH. et al. Diabetolog 42, 45 (1999)Montrose-Rafizadeh, C. et al. J Biol Chem 272, 21201 (1997)Gke R. et al. J Biol Chem 268, 19650 (1993)Eng, J. et al. J Biol Chem 267, 7402 (1992)
Catalog No. 50-210-9263