Learn More
CPC Scientific H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific EXED005B
Encompass
A new member of the Glucagon superfamily that was originally isolated from the venom of the lizard, Heloderma Horridum. At low concentrations, it interacts with putative exendin receptors causing an increase in pancreatic acinar cAMP, while at higher concentrations it interacts with VIP receptors to stimulate an increase in cellular cAMP and amylase release. ONE-LETTER SEQUENCE: HSDGTFTSDLSKQMEEEAVRLFIEWLKNGGPSSGAPPPS-NH2MOLECULAR FORMULA: C184H282N50O61S1MOLECULAR WEIGHT:4202.63STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [130357-25-4], RESEARCH AREA: DiabetesREFERENCES: J. Eng et al., J. Biol. Chem., 267, 7402 (1992)
Catalog No. 50-210-9270