Learn More
Enzo Life Sciences Exendin-4 (1mg). CAS: 141758-74-9
SDP
Catalog No. 502013904
Encompass_Preferred
Exendin-4 is a 39 aa polypeptide and incretin mimetic which was first isolated from Gila monster saliva. The synthetic form, Exenatide, is a glucagon like peptide-1 (GLP-1) agonist approved for the treatment of diabetes mellitus type II. Research shows Exendin-4 is involved in glucose-dependent enhancement of insulin secretion, reduction of hyperglycemia, glucose-dependent suppression of inappropriately high glucagon secretion, slowing of gastric emptying, and reduction of food intake, often with body weight reduction or blunting of weight gain. Alternative name: Exenatide. Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3904 Supplier Enzo Life Sciences Supplier No. ENZPRT1110001
Exendin-4 is a 39 aa polypeptide and incretin mimetic which was first isolated from Gila monster saliva. The synthetic form, Exenatide, is a glucagon like peptide-1 (GLP-1) agonist approved for the treatment of diabetes mellitus type II. Research shows Exendin-4 is involved in glucose-dependent enhancement of insulin secretion, reduction of hyperglycemia, glucose-dependent suppression of inappropriately high glucagon secretion, slowing of gastric emptying, and reduction of food intake, often with body weight reduction or blunting of weight gain. Alternative name: Exenatide. Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.