Learn More
Medchemexpress LLC HY-13443 1mg Medchemexpress, Exendin-4 CAS:141758-74-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134431MG
Encompass_Preferred
Medchemexpress, HY-13443 1mg Exendin-4 CAS:141758-74-9 Exendin-4 (Exenatide), a 39 amino acid peptide, is a long-acting glucagon-like peptide-1 receptor agonist with an IC50 of 3.22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8996