Learn More
Apexbio Technology LLC Exendin-4,20mg CAS# 141758-74-9
SDP
Supplier:  Apexbio Technology LLC A340820
Encompass_Preferred
Exendin-4, a glucagon-like protein-1 (GLP-1) receptor agonist, mimics the activity of mammalian incretin hormone glucagon-like peptide 1 (GLP-1), and thus, promotes insulin secretion and functions in the control of glucose [1].In vitro: Exendin-4 showed a pronounced effect on intracellular cAMP generation. Treatment of GLP-1 in combination with exendin-4 showed additive action on the generation of cAMP. In isolated rat islets and in mouse insulinoma beta TC-1 cells, Exendin-4 stimulated the glucose-induced insulin secretion in a dose dependent manner [2]. In basal forebrain cholinergic neurons, exendin-4 greatly reduced ibotenic acid-induced depletion of choline acetyltransferase immunoreactivity [3].Other sizes are also available. Please inqury us for quote.
Catalog No. 50-223-9985