Learn More
Bachem Exendin-4 (3-39)
SDP
Supplier:  Bachem 4027457.1000
Encompass_Preferred
Exendin-4 (3-39) , H-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2, 196109-31-6, C176H272N46O58S, Mr 3992.44, 1mg. Potent glucagon-like peptide 1 (GLP-1) receptor antagonist.
Catalog No. 50-259-782