Learn More
TARGETMOL CHEMICALS INC Exendin-4 acetate 5MG
SDP
Catalog No. 502878084
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Exendin-4 acetate is a 39 amino acid peptide.Exendin-4 acetate is a long-acting GLP-1 receptor agonist(IC50 = 3.22 nM). Purity 98.65%
Catalog No. 50-287-8084 Supplier TARGETMOL CHEMICALS INC Supplier No. T87485MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Exendin-4 acetate is a 39 amino acid peptide.Exendin-4 acetate is a long-acting GLP-1 receptor agonist(IC50 = 3.22 nM). Purity 98.65%