Learn More
Medchemexpress LLC Exendin-4 (acetate) | 914454-01-6 | MFCD08704781 | 99.9% | 4246.68 g/mol | C184H282N50O60S.C2H4O2 | 5 MG
SDP
Catalog No. 5000222025
Encompass_Preferred
Exendin-4 acetate (Exenatide acetate) is a 39-amino-acid peptide agonist of the glucagon-like peptide-1 (GLP-1) receptor, reported with an IC50 of 3.22 nM. It is provided for research use in pre-weighed quantities and is reported at high purity for laboratory applications.
  • 39-amino-acid peptide
  • Agonist of the glucagon-like peptide-1 (GLP-1) receptor
  • Reported IC50 of 3.22 nM against GLP-1 receptor
  • High purity (≈99.9%) suitable for research use
  • Available in small pre-weighed quantities (mg scale)
  • Molecular weight approximately 4,246.7 g/mol
Catalog No. 50-002-22025 Supplier Medchemexpress LLC Supplier No. HY13443A5MG
Exendin-4 acetate (Exenatide acetate) is a 39-amino-acid peptide agonist of the glucagon-like peptide-1 (GLP-1) receptor, reported with an IC50 of 3.22 nM. It is provided for research use in pre-weighed quantities and is reported at high purity for laboratory applications.
  • 39-amino-acid peptide
  • Agonist of the glucagon-like peptide-1 (GLP-1) receptor
  • Reported IC50 of 3.22 nM against GLP-1 receptor
  • High purity (≈99.9%) suitable for research use
  • Available in small pre-weighed quantities (mg scale)
  • Molecular weight approximately 4,246.7 g/mol