Learn More
Eurofins Discoverx PathHunter Exendin-4 Bioassay Kit, 10-Plate. (93-0300Y2-00030)
Supplier:  Eurofins Discoverx 930300Y200030
Encompass
The PathHunter Exendin-4 Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses Exendin-4 based activation of GLP1R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GLP1R and EA-tagged β-Arrestin-2. Activation of GLP1R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0201
Only null left
Add to Cart
Request a Quote