Learn More
Medchemexpress LLC Exendin-4 (exenatide) | 141758-74-9 | 100.0% | C184H282N50O60S | 10MG
SDP
Catalog No. 5000260082
Encompass_Preferred
Exendin-4 (exenatide) is a 39-amino-acid peptide agonist of the glucagon-like peptide-1 (GLP-1) receptor used in research to investigate GLP-1 signaling, insulin secretion, and metabolic effects. It is provided as a high-purity powder with documented sequence, potency, solubility guidance, and storage recommendations for laboratory use.
  • High receptor potency (IC50 ~3.22 nM) for GLP-1 studies.
  • Full 39-amino-acid sequence provided for experimental planning.
  • Supplied as a powder suitable for long-term storage at -80°C.
  • Soluble in DMSO and water; sonication recommended for dissolution.
  • High purity suitable for biochemical and cell-based assays.
Catalog No. 50-002-60082 Supplier Medchemexpress LLC Supplier No. HY1344310MG
Exendin-4 (exenatide) is a 39-amino-acid peptide agonist of the glucagon-like peptide-1 (GLP-1) receptor used in research to investigate GLP-1 signaling, insulin secretion, and metabolic effects. It is provided as a high-purity powder with documented sequence, potency, solubility guidance, and storage recommendations for laboratory use.
  • High receptor potency (IC50 ~3.22 nM) for GLP-1 studies.
  • Full 39-amino-acid sequence provided for experimental planning.
  • Supplied as a powder suitable for long-term storage at -80°C.
  • Soluble in DMSO and water; sonication recommended for dissolution.
  • High purity suitable for biochemical and cell-based assays.