Learn More
Genscript Corporation Exendin-4 
Supplier:  Genscript Corporation Z02811100
Encompass
Exendin-4 is a novel 39-amino acid peptide isolated from the venom of the Gila monster Heloderma suspectum. It shares 53% sequence homology with GLP-17-36amide and interacts with the same membrane receptor. Exendin-4 enhances glucose-dependent insulin secretion, suppresses inappropriately elevated glucagon secretion, and slows gastric emptying in vivo. It also promotes ß-cell proliferation and neogenesis in vitro and in animal models.
Catalog No. 50-276-606