Learn More
Enzo Life Sciences Exendin-4 monoclonal antibody (24) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDABS012241
Encompass_Preferred
Clone: 24. Isotype: IgG1κ. Immunogen: Synthetic exendin-4 from Gila monster Heloderma suspectum. Species reactivity: Gila monster (Heloderma suspectum). Specificity: Recognizes Gila monster Heloderma suspectum exendin-4 which has a high degree of homology with GLP-1 found in mammals. Applications: ELISA. Purity: Protein A/G-affinity purified. Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.5M sodium chloride and 15mM sodium azide.
Catalog No. 50-201-2390