Learn More
Pyramex™ Exeter™ Safety Eyewear
Catalog No. 19155746 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Charcoal Gray
Gloss Black
Lens Tint:
Amber
Blue, Mirror
Clear
Gray
Indoor/Outdoor, Mirror
Red
Silver, Mirror
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-155-746 Gloss Black Clear
19-155-747 Gloss Black Gray
19-155-748 Gloss Black Amber
19-155-749 Gloss Black Red
19-155-751 Gloss Black Blue, Mirror
19-155-750 Gloss Black Silver, Mirror
19-155-752 Gloss Black Indoor/Outdoor, Mirror
19-155-922 Charcoal Gray Blue, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-155-746 Supplier Pyramex™ Supplier No. SB5110DT
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Wrap-around, sunglass style fits all facial sizes
  • Demonstrates strength with style and protection
  • Sporty style with maximum protection
  • Curved lens with side window gives fashionable look with panoramic view
  • Durable frame gives a pressure free fit
  • Soft nose piece and rubber insert temple tips provide a comfortable fit
  • Scratch resistant lenses filter out 99.9% of harmful UV rays
  • Specifications
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Gloss Black
    Weight (Metric) 29 g
    Lens Material Polycarbonate
    Frame Material Nylon