Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Exhaust Fan 16In Bl Fiberglass

Catalog No. 502793786
Encompass_Preferred

Exhaust Fan 16In Bl Fiberglass 115V

Catalog No. 50-279-3786 Supplier Grainger Supplier No. 1BLJ1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Exhaust Fan 16In Bl Fiberglass 115V

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.