Learn More
Chemglass Life Sciences Exhaust Gas Outlet Column
SDP
Catalog No. 501531252
Encompass_Preferred
Exhaust Gas Outlet Column. Heavy wall column, 4.5 long having a sidearm with a 24/40 outer joint. The 24/40 outer joint can be used with an adapter for connection to vacuum source. Adapter has an overall width of 7.
Catalog No. 50-153-1252 Supplier Chemglass Life Sciences Supplier No. CG183015
Exhaust Gas Outlet Column. Heavy wall column, 4.5 long having a sidearm with a 24/40 outer joint. The 24/40 outer joint can be used with an adapter for connection to vacuum source. Adapter has an overall width of 7.