Learn More
Chemglass Life Sciences Chemglass Exhaust Gas Outlet for 20 L Gas Scrubber, 100 mm Column (CG-1830-54)
SDP
Catalog No. 502733577
Encompass_Preferred
Exhaust gas outlet designed for use with a 20 L gas scrubber fitted with a 100 mm column. It provides a controlled exhaust connection for laboratory gas-scrubbing systems and is intended as a replacement or spare component for compatible scrubber assemblies.
  • Provides controlled exhaust connection.
  • Fits 100 mm column on 20 L scrubber.
  • Made from laboratory-grade glass for chemical resistance.
  • Sold individually for replacement or repair.
Catalog No. 50-273-3577 Supplier Chemglass Life Sciences Supplier No. CG183054
Add to Cart
Add to Cart
Exhaust gas outlet designed for use with a 20 L gas scrubber fitted with a 100 mm column. It provides a controlled exhaust connection for laboratory gas-scrubbing systems and is intended as a replacement or spare component for compatible scrubber assemblies.
  • Provides controlled exhaust connection.
  • Fits 100 mm column on 20 L scrubber.
  • Made from laboratory-grade glass for chemical resistance.
  • Sold individually for replacement or repair.