Learn More
MSC Guardair Exhaust Housing, with Silencer Foam, 88 dBA Sound Reduction
Catalog No. 502836538
Encompass
This vented exhaust housing with silencer foam is designed to significantly reduce the operational noise of compatible vacuums, bringing the sound level down to 88 dBA or less. It features dimensions of 5.8 in L x 8.3 in W x 8.1 in H and weighs 2.7 lb. Each unit includes an exhaust housing, three mounting screws, and one exhaust silencer foam insert for easy installation and immediate noise reduction.
  • Vented exhaust housing with silencer foam
  • Reduces sound level to 88 dBA or less
  • Designed for specific vacuum types
  • Compatible with drum top vacuums
  • Includes exhaust housing, mounting screws, and silencer foam inserts
Catalog No. 50-283-6538 Supplier MSC Supplier No. 64361629
This vented exhaust housing with silencer foam is designed to significantly reduce the operational noise of compatible vacuums, bringing the sound level down to 88 dBA or less. It features dimensions of 5.8 in L x 8.3 in W x 8.1 in H and weighs 2.7 lb. Each unit includes an exhaust housing, three mounting screws, and one exhaust silencer foam insert for easy installation and immediate noise reduction.
  • Vented exhaust housing with silencer foam
  • Reduces sound level to 88 dBA or less
  • Designed for specific vacuum types
  • Compatible with drum top vacuums
  • Includes exhaust housing, mounting screws, and silencer foam inserts