Learn More
Grainger Fantech Exterior Wall Mount Centrifugal Exhaust Fan, 6 in. Duct, 242 CFM, Galvanized Steel, 120 V
Catalog No. 502760918
Encompass_Preferred
The RVF Series centrifugal exhaust fan is designed for exterior wall-mount installation to remotely extract stale, humid, or polluted air from bathrooms, kitchens, utility rooms, and garages. Motor noise is effectively kept outside the building, ensuring quiet indoor operation. This lightweight, compact fan features a durable galvanized sheet metal housing with a white powder-coat finish and utilizes backward-curved impeller blades for efficient airflow performance.
  • Backward-curved impeller engineered for high efficiency and minimized noise
  • Automatic reset thermal overload protection safeguards the motor
  • IP44-rated external-rotor motor provides protection against dust and moisture
  • Rubber gasket seal enables tight, leak-free duct connections
  • Suitable for continuous operation in air temperatures up to 140 °F
Catalog No. 50-276-0918 Supplier Grainger Supplier No. 15W795
The RVF Series centrifugal exhaust fan is designed for exterior wall-mount installation to remotely extract stale, humid, or polluted air from bathrooms, kitchens, utility rooms, and garages. Motor noise is effectively kept outside the building, ensuring quiet indoor operation. This lightweight, compact fan features a durable galvanized sheet metal housing with a white powder-coat finish and utilizes backward-curved impeller blades for efficient airflow performance.
  • Backward-curved impeller engineered for high efficiency and minimized noise
  • Automatic reset thermal overload protection safeguards the motor
  • IP44-rated external-rotor motor provides protection against dust and moisture
  • Rubber gasket seal enables tight, leak-free duct connections
  • Suitable for continuous operation in air temperatures up to 140 °F