Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ EXI-410-FL Inverted Microscope Series Trinocular with Plan Phase Objectives and FITC/GFP Fluorescence
SDP

Catalog No. 01189195 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Head Style:
Trinocular
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-189-195 Trinocular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-189-195 Supplier ACCU-SCOPE™ Supplier No. EXI410FL470
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The ACCU-SCOPE™EXI-410-FL-470 inverted microscope with FITC/GFP Fluorescence delivers outstanding optical performance, convenience, flexibility and value to researchers, educators, and clinicians. Ideal for the daily rigors of tissue culture applications and more advanced observation methods.

  • EXI-410 Inverted Fluorescence Microscope
  • Trinocular viewing head, inclined 45°, with attachable side mounted camera port
  • Trinocular viewing head can be rotated to raise/lower eyetubes by 35 mm
  • Eyepiece/Camera Port: 100/0 or 0/100
  • WF10x/22 mm eyepieces with roll down eyeguards
  • Rear-facing quintuple nosepiece
  • Infinity corrected plan4x, Plan Phase 10x and 20x objectives
  • Additional Objectives, and filter sets, LED Episcopic components can be added (up to two additional)
  • Fixed plain stage, 170 x 250 mm with metal and glass insert plates
  • ELWD condenser, N.A. 0.3, working distance = 75 mm
  • Variable LED diascopic illuminator
  • Dust cover, power cord, Universal power supply 100 to 240 V
  • Phase Slider with 10x/20x/40x phase annulli, phase centering telescope
  • FITC/GFP filter set for EXI-410 Series, EX: 470/40x, BS: 505DC, EM: 535/50m
  • Epi-fluorescence illumination configurable with up to 3 different fluorescence LED units
  • Epi-fluorescence filter turret with 3 filter cube positions
  • Contrast shield for enhanced fluorescence illumination

Specifications

Voltage 100/240 V
Magnification Power 4X, 10X, 20X
Objective Types Infinity Corrected Plan Achromatic, Phase Contrast
Eyepiece WF 10X/22 mm
Lighting Type LED
Head Style Trinocular
Camera Type None
Display Type None
Model EXI-410
Product Type Inverted Phase-contrast Fluorescent Microscope
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.