Learn More
Grainger Exit Sign 1.0W Red/Green 1 or 2 Faces
Catalog No. 502765247
Encompass_Preferred
The Cooper Lighting Exit Sign Emergency Battery Pack is a self-contained emergency lighting unit designed for exit signs. It contains a sealed lead acid battery that provides 90 minutes of emergency power. The unit has a solid-state charger and test switch and provides 6V or 12V output at 87.5 lumens. It is made of steel with a white powder-coated finish and is UL listed. The dimensions are 9-3/4" x 6-5/8" x 2-3/8". This emergency battery pack provides reliable power to illuminate exit signs in the event of an outage.
Catalog No. 50-276-5247 Supplier Grainger Supplier No. 16U375
The Cooper Lighting Exit Sign Emergency Battery Pack is a self-contained emergency lighting unit designed for exit signs. It contains a sealed lead acid battery that provides 90 minutes of emergency power. The unit has a solid-state charger and test switch and provides 6V or 12V output at 87.5 lumens. It is made of steel with a white powder-coated finish and is UL listed. The dimensions are 9-3/4" x 6-5/8" x 2-3/8". This emergency battery pack provides reliable power to illuminate exit signs in the event of an outage.