Learn More
Grainger Exit Sign Less Than 1.0W Red 1 or 2
Catalog No. 502764873
Encompass_Preferred
The Grainger lighted exit signs are designed to clearly indicate emergency exits in commercial buildings. They feature durable steel or aluminum housings and long-lasting LED lamps that provide bright illumination and energy efficiency. The signs come in single or double face configurations with universal mounting for easy installation. They are available with a variety of legends including exit, emergency exit, and running man. These signs meet building and fire codes and are UL listed for safety. With a 5 year warranty, these high quality signs ensure your building's exits are clearly marked.
Catalog No. 50-276-4873 Supplier Grainger Supplier No. 16U370
The Grainger lighted exit signs are designed to clearly indicate emergency exits in commercial buildings. They feature durable steel or aluminum housings and long-lasting LED lamps that provide bright illumination and energy efficiency. The signs come in single or double face configurations with universal mounting for easy installation. They are available with a variety of legends including exit, emergency exit, and running man. These signs meet building and fire codes and are UL listed for safety. With a 5 year warranty, these high quality signs ensure your building's exits are clearly marked.