Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

exo-2-Norbornyl Formate 99%, Thermo Scientific™

Catalog No. AC185550500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC185550500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC185550500 Supplier Thermo Scientific Chemicals Supplier No. 185550500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41498-71-9
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
InChI Key SGXIEZNAOCVSKO-KAVNDROISA-N
Synonym exo-2-norbornyl formate, 1r,2r,4s-bicyclo 2.2.1 heptan-2-yl formate, formic acid 1alpha,4alpha-bicyclo 2.2.1 heptan-2alpha-yl ester
PubChem CID 133126528
IUPAC Name [(3R)-3-bicyclo[2.2.1]heptanyl] formate
SMILES C1CC2CC1CC2OC=O

Specifications

Density 1.04
Quantity 50 g
Formula Weight 140.18
Chemical Name or Material exo-2-Norbornyl formate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.