Learn More
Medchemexpress LLC exo-BCN-NHS carbonat 25mg
SDP
Catalog No. 5000204561
Encompass_Preferred
exo-BCN-NHS carbonate carbonate contains BCN group Strain-promoted alkyne-azide cycloaddition (SPAAC) can also occur with molecules containing DBCO or BCN groups
Catalog No. 50-002-04561 Supplier Medchemexpress LLC Supplier No. 25MGHYW096149
exo-BCN-NHS carbonate carbonate contains BCN group Strain-promoted alkyne-azide cycloaddition (SPAAC) can also occur with molecules containing DBCO or BCN groups