Learn More
Medchemexpress LLC Exo-BCN-PEG7-NHCO-maleimide | 2143965-47-1 | 95.1% | C34H53N3O12 | 10MG
SDP
Catalog No. 5000260650
Encompass_Preferred
BCN-exo-PEG7-maleimide is an ADC linker and bioconjugation reagent that combines a strain-promoted bicyclononyne (BCN) for copper-free azide-alkyne cycloaddition with a maleimide group for thiol-specific coupling, connected by a PEG7 spacer to enhance solubility and linker flexibility.
  • Enables copper-free click chemistry with azide-containing partners.
  • Provides thiol-specific conjugation via a maleimide group.
  • PEG7 spacer improves solubility and linker flexibility.
  • Suitable for antibody-drug conjugate synthesis and other bioconjugation workflows.
  • Available with documented purity and storage instructions for reproducible use.
Catalog No. 50-002-60650 Supplier Medchemexpress LLC Supplier No. HY13473410MG
BCN-exo-PEG7-maleimide is an ADC linker and bioconjugation reagent that combines a strain-promoted bicyclononyne (BCN) for copper-free azide-alkyne cycloaddition with a maleimide group for thiol-specific coupling, connected by a PEG7 spacer to enhance solubility and linker flexibility.
  • Enables copper-free click chemistry with azide-containing partners.
  • Provides thiol-specific conjugation via a maleimide group.
  • PEG7 spacer improves solubility and linker flexibility.
  • Suitable for antibody-drug conjugate synthesis and other bioconjugation workflows.
  • Available with documented purity and storage instructions for reproducible use.