Learn More
System Biosciences LLC EXO-DEPLETED HEAT INACTIVE FBSExosome-depleted FBS Media Supplement Heat Inactivated = 250 ml
SDP
Catalog No. NC0573808
Encompass_Preferred
Bovine exosome-free FBS that has been heat inactivated, for labs that would like to use Exo-FBS but require heat inactivation. Fetal bovine serum, or FBS, is an important component of many types of cell culture media, and some researchers require heat inactivated FBS. But for researchers interested in isolating exosomes from cultured cells, standard heat inactivated FBS can introduce unwanted complications—bovine exosomes, which can cause significant background issues or interfere with functional studies. Which is why SBI developed heat inactivated Exo-FBS, our patented exosome-depleted FBS.
Catalog No. NC0573808 Supplier System Biosciences LLC Supplier No. EXOFBSHI250A1
Bovine exosome-free FBS that has been heat inactivated, for labs that would like to use Exo-FBS but require heat inactivation. Fetal bovine serum, or FBS, is an important component of many types of cell culture media, and some researchers require heat inactivated FBS. But for researchers interested in isolating exosomes from cultured cells, standard heat inactivated FBS can introduce unwanted complications—bovine exosomes, which can cause significant background issues or interfere with functional studies. Which is why SBI developed heat inactivated Exo-FBS, our patented exosome-depleted FBS.