Learn More
System Biosciences LLC Exo-Fect™ Exosome Transfection KitExo-Fect Exosome Transfection Kit = 10 Reactions
SDP
Catalog No. NC0733644
Encompass_Preferred
Turn your exosomes into delivery vehicles with Exo-Fect—enables insertion of RNAs, DNAs (including plasmids), and small molecules into isolated exosomes. With Exo-Fect, you can turn isolated exosomes into cargo delivery vehicles that introduce RNAs, DNAs (including plasmids), and even small molecules into recipient cells. Simply combine isolated exosomes with Exo-Fect and your desired cargo, and in less than an hour your loaded exosomes will be ready to add to recipient cells.
Catalog No. NC0733644 Supplier System Biosciences LLC Supplier No. EXFT10A1
Turn your exosomes into delivery vehicles with Exo-Fect—enables insertion of RNAs, DNAs (including plasmids), and small molecules into isolated exosomes. With Exo-Fect, you can turn isolated exosomes into cargo delivery vehicles that introduce RNAs, DNAs (including plasmids), and even small molecules into recipient cells. Simply combine isolated exosomes with Exo-Fect and your desired cargo, and in less than an hour your loaded exosomes will be ready to add to recipient cells.