Learn More
System Biosciences LLC EXOGLOW LABELING KITExoGlow™-Vivo EV Labeling Kit (Near IR) = 12 Reactions
SDP
Catalog No. NC1765001
Encompass_Preferred
Monitor EV trafficking in vivo with the ExoGlow-Vivo EV Labeling Kit (Near IR), a unique dye developed for following Exosomes in animal models and tissues. Visualizing extracellular vesicles (EVs) in cells and organisms has been technically difficult due to the high background from auto-fluorescence in the UV and visible areas of the spectrum and a lack of specificity from the typically lipophilic dyes. SBI’s ExoGlow-Vivo EV Labeling Kit (Near IR) is the first reagent specifically designed to overcome these problems through the use of a proprietary, non-lipophilic dye that emits in the near infrared (NIR) range (excitation at 784 nm; emission at 806 nm). Delivering a level of specificity and sensitivity that takes the guesswork out of tracking EVs in vivo, ExoGlow-Vivo is ideal for EV biodistribution and kinetic studies needed to fully realize the value of EVs in basic research and translational applications.
Catalog No. NC1765001 Supplier System Biosciences LLC Supplier No. EXOGV900A1
Monitor EV trafficking in vivo with the ExoGlow-Vivo EV Labeling Kit (Near IR), a unique dye developed for following Exosomes in animal models and tissues. Visualizing extracellular vesicles (EVs) in cells and organisms has been technically difficult due to the high background from auto-fluorescence in the UV and visible areas of the spectrum and a lack of specificity from the typically lipophilic dyes. SBI’s ExoGlow-Vivo EV Labeling Kit (Near IR) is the first reagent specifically designed to overcome these problems through the use of a proprietary, non-lipophilic dye that emits in the near infrared (NIR) range (excitation at 784 nm; emission at 806 nm). Delivering a level of specificity and sensitivity that takes the guesswork out of tracking EVs in vivo, ExoGlow-Vivo is ideal for EV biodistribution and kinetic studies needed to fully realize the value of EVs in basic research and translational applications.